Assay Details
Target Gene Details
Entrez Gene ID: | 59054 |
Gene Name: | mitochondrial ribosomal protein S30 |
Gene Aliases: |
2610020A16Rik, AA968347, PAP, Pdcd9 |
Location: |
Chr.13:118380110-118387252 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps30 | NM_021556.3 | 5 | 1399 | NP_067531.1 |
| AK011476.1 | 5 | 1401 | BAB27644.1 | |
| AK013360.1 | ||||
| AK151642.1 | 5 | 1400 | BAE30572.1 | |
| AL355719.1 | CAB90812.1 | |||
| BC029784.1 | AAH29784.1 |
More Information
Set Membership: |
|

Genomic Map