Assay Details
Target Gene Details
Entrez Gene ID: | 105245 |
Gene Name: | thioredoxin domain containing 5 |
Gene Aliases: |
AL022641, ERp46, PC-TRP |
Location: |
Chr.13:38500266-38528824 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
More Information
Set Membership: |
|

Genomic Map