Assay Details
Target Gene Details
Entrez Gene ID: | 66864 |
Gene Name: | C-type lectin domain family 14, member a |
Gene Aliases: |
1200003C23Rik, AI642649 |
Location: |
Chr.12:58264720-58269258 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Clec14a | NM_025809.5 | 1 | 1516 | NP_080085.3 |
AK004557.2 | 1 | 1156 | BAB23370.2 | |
AK014681.1 | 1 | 1462 | BAB29502.1 | |
AK018432.1 | 1 | 1130 | BAB31209.1 | |
AK033733.1 | 1 | 1193 | BAC28454.1 | |
AK045596.1 | 1 | 1147 | BAC32430.1 | |
AK080201.1 | 1 | 1130 | BAC37846.1 | |
AK164665.1 | 1 | 1135 | BAE37865.1 | |
BC019452.1 | 1 | 1443 | AAH19452.1 |
