Assay Details
Target Gene Details
Entrez Gene ID: | 21869 |
Gene Name: | NK2 homeobox 1 |
Gene Aliases: |
AV026640, Nkx2.1, T/EBP, Titf1, Ttf-1 |
Location: |
Chr.12:56531935-56536908 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nkx2-1 | NM_009385.3 | 1 | 506 | NP_033411.3 |
| BC057607.1 | 1 | 506 | AAH57607.1 |
More Information
Set Membership: |
|

Genomic Map