Assay Details
Target Gene Details
Entrez Gene ID: | 319163 |
Gene Name: | histone cluster 1, H2aa |
Gene Aliases: |
Th2a |
Location: |
Chr.13:23934462-23934913 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hist1h2aa | NM_175658.2 | 1 | 132 | NP_783589.1 |
| BC107354.1 | 1 | 132 | AAI07355.1 | |
| BC107355.1 | 1 | 132 | AAI07356.1 |
More Information
Set Membership: |
|

Genomic Map