Assay Details
Target Gene Details
Entrez Gene ID: | 22169 |
Gene Name: | cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial |
Gene Aliases: |
1200004E04Rik, TDKI, Tyki |
Location: |
Chr.12:26469059-26479837 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmpk2 | NM_020557.4 | 5 | 2276 | NP_065582.3 |
| AK004595.1 | 5 | 2111 | BAB23396.1 | |
| AK150512.1 | 5 | 2101 | BAE29625.1 | |
| AK150543.1 | 5 | 2103 | BAE29646.1 | |
| AK150725.1 | 5 | 2101 | BAE29803.1 | |
| AK150885.1 | 5 | 2103 | BAE29932.1 | |
| AK153429.1 | BAE31987.1 | |||
| AK153469.1 | 5 | 2108 | BAE32020.1 | |
| BC027329.1 | 5 | 2175 | AAH27329.1 | |
| BC057565.1 | 5 | 2089 | AAH57565.1 | |
| L32973.1 | 4 | 2361 | AAA58770.1 |
More Information
Set Membership: |
|

Genomic Map