Assay Details
Target Gene Details
Entrez Gene ID: | 211064 |
Gene Name: | alkB, alkylation repair homolog 1 (E. coli) |
Gene Aliases: |
2700073G19Rik, Abh, Alkbh, alkB, hABH |
Location: |
Chr.12:87428077-87443839 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alkbh1 | NM_001102565.1 | NP_001096035.1 | ||
| AK042102.1 | ||||
| AK143105.1 | ||||
| AK144828.1 |
Target Gene Details
Entrez Gene ID: | 654309 |
Gene Name: | neural regeneration protein |
Gene Aliases: |
- |
Location: |
Chr.12:87442848-87444017 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nrp | NM_001013372.2 | 1 | 203 | NP_001013390.1 |
| AK012518.1 | ||||
| AK049549.1 | BAC33805.1 | |||
| AY753183.1 | 1 | 203 | AAU93563.1 | |
| BC119399.1 | 1 | 180 | ||
| BC119401.1 | 1 | 180 |
More Information
Set Membership: |
|

Genomic Map