Assay Details
Target Gene Details
Entrez Gene ID: | 70881 |
Gene Name: | 5'-nucleotidase, cytosolic IB |
Gene Aliases: |
4921514H13Rik, AIRP, CN-IB |
Location: |
Chr.12:10369971-10390175 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nt5c1b | NM_027588.3 | 4 | 700 | NP_081864.2 |
| AF356186.1 | 4 | 667 | AAK39109.1 | |
| AJ295253.1 | 4 | 627 | CAC44364.1 | |
| AK014892.1 | 4 | 700 | ||
| BC060976.1 | 4 | 686 | AAH60976.1 |

Genomic Map