Assay Details
Target Gene Details
Entrez Gene ID: | 627191 |
Gene Name: | synapse differentiation inducing 1 like |
Gene Aliases: |
DSPC1, Gm261, Tmem90a |
Location: |
Chr.12:84677277-84698907 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syndig1l | NM_001033334.2 | 4 | 1967 | NP_001028506.1 |
| AK140195.1 | 4 | 1967 | BAE24274.1 |

Genomic Map