Assay Details
Target Gene Details
Entrez Gene ID: | 319177 |
Gene Name: | histone cluster 1, H2ba |
Gene Aliases: |
Th2b |
Location: |
Chr.13:23933725-23934156 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hist1h2ba | NM_175663.2 | 1 | 58 | NP_783594.1 |
| BC126923.1 | 1 | 58 |
More Information
Set Membership: |
|

Genomic Map