Assay Details
Target Gene Details
Entrez Gene ID: | 66336 |
Gene Name: | centromere protein P |
Gene Aliases: |
1700022C02Rik, 4921518G09Rik |
Location: |
Chr.13:49464025-49652879 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cenpp | NM_025495.3 | NP_079771.1 | ||
| AK006234.1 | BAB24472.1 | |||
| AK012863.1 | BAB28519.1 | |||
| BC147207.1 | ||||
| BC147208.1 |
Target Gene Details
Entrez Gene ID: | 407800 |
Gene Name: | extracellular matrix protein 2, female organ and adipocyte specific |
Gene Aliases: |
9030618O22Rik, BC065151, tenonectin |
Location: |
Chr.13:49504810-49532789 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ecm2 | NM_001012324.2 | 4 | 1098 | NP_001012324.1 |
| AK038812.1 | 4 | 960 | BAE20541.1 | |
| AK040995.1 | 4 | 1122 | BAE20587.1 | |
| AK134746.1 | 4 | 1052 | BAE22265.1 | |
| AK163501.1 | 3 | 3120 | ||
| BC065151.1 | 4 | 968 | AAH65151.1 | |
| BC089559.1 | 4 | 1098 | AAH89559.1 |
More Information
Set Membership: |
|

Genomic Map