Assay Details
Target Gene Details
Entrez Gene ID: | 76485 |
Gene Name: | glycosyltransferase 8 domain containing 1 |
Gene Aliases: |
2410004H05Rik, 5430414N14Rik, AI450005 |
Location: |
Chr.14:31001347-31012441 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glt8d1 | NM_001165930.1 | 10 | 1332 | NP_001159402.1 |
| NM_029626.2 | 10 | 1861 | NP_083902.2 | |
| AK010388.1 | 10 | 1590 | BAB26903.1 | |
| AK077388.1 | 10 | 1325 | ||
| AK208639.1 | 1 | 203 | ||
| AK211086.1 | 1 | 199 | ||
| BC055709.1 | 9 | 1748 | AAH55709.1 | |
| BC058731.1 | 9 | 1748 | AAH58731.1 | |
| BC069873.1 | 10 | 1411 | AAH69873.1 |
More Information
Set Membership: |
|

Genomic Map