Assay Details
Target Gene Details
Entrez Gene ID: | 105239 |
Gene Name: | ring finger protein 44 |
Gene Aliases: |
AI854545, mKIAA1100 |
Location: |
Chr.13:54679399-54693995 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnf44 | NM_001146025.1 | 11 | 2399 | NP_001139497.1 |
| NM_001146026.1 | 11 | 2324 | NP_001139498.1 | |
| NM_001146027.1 | 11 | 2190 | NP_001139499.1 | |
| NM_134064.2 | 11 | 2332 | NP_598825.2 | |
| NR_027395.1 | 11 | 2252 | ||
| NR_027396.1 | 11 | 2249 | ||
| AK147349.1 | 11 | 2325 | BAE27859.1 | |
| AK160090.1 | 11 | 2183 | BAE35621.1 | |
| AK160115.1 | 11 | 2183 | BAE35639.1 | |
| AK161651.1 | 11 | 2314 | ||
| AK171943.1 | 11 | 2181 | BAE42740.1 | |
| AK172257.1 | 11 | 2182 | ||
| BC017630.1 | 11 | 2125 | AAH17630.1 | |
| BC035548.1 | 11 | 2147 | AAH35548.1 |

Genomic Map