Assay Details
Target Gene Details
Entrez Gene ID: | 105835 |
Gene Name: | small G protein signaling modulator 3 |
Gene Aliases: |
1810012I01Rik, AI428557, BB175482, Cip85, R75178, RUSC3, Rutbc3 |
Location: |
Chr.15:80977765-81012290 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgsm3 | NM_134091.2 | 22 | 2888 | NP_598852.2 |
| AI844026.1 | 1 | 126 | ||
| AK142314.1 | 22 | 2856 |
More Information
Set Membership: |
|

Genomic Map