Assay Details
Target Gene Details
Entrez Gene ID: | 75501 |
Gene Name: | RIKEN cDNA 1700010K23 gene |
Gene Aliases: |
- |
Location: |
Chr.16:66657118-66664626 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700010K23Rik | NR_040512.1 | |||
| AK005838.1 | ||||
| BY705986.1 |
Target Gene Details
Entrez Gene ID: | 239857 |
Gene Name: | cell adhesion molecule 2 |
Gene Aliases: |
2900078E11Rik, 9330131D06, A830029E02Rik, Igdf4d, Igsf4d, NECL3, SynCAM 2, SynCAM2 |
Location: |
Chr.16:66655421-67621200 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadm2 | NM_001145977.1 | 9 | 6940 | NP_001139449.1 |
| NM_178721.4 | 10 | 6967 | NP_848836.2 | |
| AK046773.1 | 1 | 928 | ||
| AK047633.1 | 1 | 1829 |
More Information
Set Membership: |
|

Genomic Map