Assay Details
Target Gene Details
Entrez Gene ID: | 75288 |
Gene Name: | solute carrier family 35, member F4 |
Gene Aliases: |
4930550L21Rik |
Location: |
Chr.14:49298519-49525837 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35f4 | NM_029238.2 | 8 | 1518 | NP_083514.1 |
| AB041538.1 | 9 | 1446 | BAA95023.1 | |
| AK034308.1 | 8 | 1520 | BAC28670.1 | |
| BC138981.1 | 8 | 1417 | ||
| BC145720.1 | 8 | 1417 |

Genomic Map