Assay Details
Target Gene Details
Entrez Gene ID: | 69906 |
Gene Name: | solute carrier family 25, member 32 |
Gene Aliases: |
2610043O12Rik, Mftc |
Location: |
Chr.15:39094191-39112768 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a32 | NM_172402.3 | 1 | 248 | NP_765990.2 |
| AK031187.1 | 1 | 53 | BAC27295.1 | |
| AK145178.1 | 1 | 208 | BAE26277.1 | |
| AK167501.1 | 1 | 58 | BAE39577.1 | |
| AK170643.1 | 1 | 217 | BAE41931.1 | |
| AK172029.1 | 1 | 216 | BAE42785.1 | |
| AK172597.1 | 1 | 206 | BAE43085.1 | |
| BB641480.1 | 1 | 249 | ||
| BC031874.1 | 1 | 182 | AAH31874.1 |
More Information
Set Membership: |
|

Genomic Map