Assay Details
Target Gene Details
Entrez Gene ID: | 94352 |
Gene Name: | lysyl oxidase-like 2 |
Gene Aliases: |
1110004B06Rik, 4930526G11Rik, 9430067E15Rik |
Location: |
Chr.14:69608692-69695834 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Loxl2 | NM_033325.2 | 14 | 4708 | NP_201582.2 |
| AK003401.1 | 1 | 57 |
More Information
Set Membership: |
|

Genomic Map