Assay Details
Target Gene Details
Entrez Gene ID: | 68453 |
Gene Name: | GPI-anchored HDL-binding protein 1 |
Gene Aliases: |
1110002J19Rik, GPI-HBP1 |
Location: |
Chr.15:75596658-75598213 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpihbp1 | NM_026730.1 | 4 | 397 | NP_081006.1 |
| AB095543.1 | 4 | 404 | BAC23061.1 | |
| AK003305.1 | 4 | 413 | BAB22704.1 | |
| BC061225.1 | 4 | 397 | AAH61225.1 |

Genomic Map