Assay Details
Target Gene Details
Entrez Gene ID: | 18693 |
Gene Name: | protein interacting with C kinase 1 |
Gene Aliases: |
Prkcabp |
Location: |
Chr.15:79229166-79249466 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pick1 | NM_001045558.1 | 13 | 1598 | NP_001039023.1 |
| NM_008837.3 | 13 | 1657 | NP_032863.2 | |
| AI385692.1 | 1 | 387 | ||
| AI893973.1 | 2 | 421 | ||
| BC048788.1 | 13 | 1543 | AAH48788.1 | |
| Z46720.1 | 12 | 1346 | CAA86675.1 |
More Information
Set Membership: |
|

Genomic Map