Assay Details
Target Gene Details
Entrez Gene ID: | 223697 |
Gene Name: | Sad1 and UNC84 domain containing 2 |
Gene Aliases: |
B230369L08Rik, C030011B15, Unc84b |
Location: |
Chr.15:79724068-79742536 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sun2 | NM_001205345.1 | 18 | 3065 | NP_001192274.1 |
| NM_001205346.1 | 18 | 3059 | NP_001192275.1 | |
| NM_194342.3 | 17 | 2969 | NP_919323.2 | |
| AK046318.1 | ||||
| AK047688.1 | 6 | 1411 | ||
| AK080116.1 | 2 | 798 | BAC37829.1 | |
| AK128958.1 | 17 | 2963 | BAC87662.1 | |
| AK156246.1 | 18 | 3066 | BAE33640.1 | |
| AK171058.1 | 18 | 3028 | BAE42217.1 | |
| AK172398.1 | 18 | 3067 | ||
| AK172996.1 | 16 | 3669 | BAD32274.1 | |
| BC098208.1 | 17 | 3310 | AAH98208.1 |
More Information
Set Membership: |
|

Genomic Map