Assay Details
Target Gene Details
Entrez Gene ID: | 93834 |
Gene Name: | pellino 2 |
Gene Aliases: |
AW047589, BB129927 |
Location: |
Chr.14:48120778-48308103 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Peli2 | NM_033602.2 | 6 | 5041 | NP_291080.2 |
| AK079187.1 | 1 | 839 |
More Information
Set Membership: |
|

Genomic Map