Assay Details
Target Gene Details
Entrez Gene ID: | 84544 |
Gene Name: | CD96 antigen |
Gene Aliases: |
1700109I12Rik, Tactile |
Location: |
Chr.16:46035657-46120248 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd96 | NM_032465.2 | 15 | 1970 | NP_115854.2 |
| AK007152.1 | 3 | 286 | ||
| AK040539.1 | 15 | 1974 | BAC30620.1 | |
| AK156470.1 | 15 | 1971 | BAE33723.1 | |
| AY029156.1 | 15 | 1828 | AAK31788.1 | |
| BC052865.1 | 15 | 1867 | AAH52865.1 |
More Information
Set Membership: |
|

Genomic Map