Assay Details
Target Gene Details
Entrez Gene ID: | 320873 |
Gene Name: | cadherin 10 |
Gene Aliases: |
A830016G23Rik, C030003B10Rik, C030011H18Rik |
Location: |
Chr.15:18818888-19016206 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh10 | NM_009865.2 | 12 | 2711 | NP_033995.1 |
| AF183946.1 | 12 | 2480 | AAL67951.1 | |
| AK138606.1 | 12 | 2713 | BAE23714.1 | |
| AK161927.1 | 1 | 1753 | ||
| BC052056.1 | 12 | 2419 | AAH52056.1 | |
| BC055808.1 | 3 | 1282 | AAH55808.1 | |
| BC062962.1 | 12 | 2419 | AAH62962.1 | |
| U69137.1 | 3 | 675 | AAB87708.1 |
More Information
Set Membership: |
|

Genomic Map