Assay Details
Target Gene Details
Entrez Gene ID: | 432940 |
Gene Name: | OTU deubiquitinase with linear linkage specificity |
Gene Aliases: |
C79097, Fam105b, m3Sapc, m7-1Sapc |
Location: |
Chr.15:27605917-27630980 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Otulin | NM_001013792.2 | 7 | 1128 | NP_001013814.2 |
| AK134955.1 | 7 | 963 | BAE22354.1 | |
| AK150371.1 | 7 | 1131 | BAE29504.1 | |
| BC028541.1 | 2 | 328 | AAH28541.1 | |
| BC087945.1 | 4 | 729 | AAH87945.1 |

Genomic Map