Assay Details
Target Gene Details
Entrez Gene ID: | 73225 |
Gene Name: | family with sequence similarity 118, member A |
Gene Aliases: |
- |
Location: |
Chr.15:85037015-85116933 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam118a | NM_133750.4 | 6 | 981 | NP_598511.1 |
| AK143690.1 | 6 | 954 | BAE25500.1 | |
| BC016425.1 | 6 | 839 | AAH16425.1 | |
| BC058794.1 | 6 | 1042 | AAH58794.1 | |
| BC066174.1 | 6 | 897 | AAH66174.1 |

Genomic Map