Assay Details
Target Gene Details
Entrez Gene ID: | 223701 |
Gene Name: | MKL (megakaryoblastic leukemia)/myocardin-like 1 |
Gene Aliases: |
AMKL, Bsac, Mal, Mrtf-A |
Location: |
Chr.15:81012281-81190757 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mkl1 | NM_001082536.1 | 14 | 3097 | NP_001076005.1 |
| NM_153049.3 | 13 | 2859 | NP_694629.2 | |
| AF385582.1 | 13 | 2630 | AAM94258.1 | |
| AF532597.1 | 14 | 2981 | AAN33041.1 | |
| AK044188.1 | 14 | 3090 | BAC31809.1 | |
| AK155821.1 | 14 | 3103 | ||
| AK155937.1 | 13 | 2843 | BAE33511.1 | |
| BC050941.1 | 14 | 2990 | AAH50941.1 |
More Information
Set Membership: |
|

Genomic Map