Assay Details
Target Gene Details
Entrez Gene ID: | 223701 |
Gene Name: | MKL (megakaryoblastic leukemia)/myocardin-like 1 |
Gene Aliases: |
AMKL, Bsac, Mal, Mrtf-A |
Location: |
Chr.15:81012281-81190757 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mkl1 | NM_001082536.1 | 9 | 1253 | NP_001076005.1 |
| NM_153049.3 | 8 | 1015 | NP_694629.2 | |
| AF385582.1 | 8 | 786 | AAM94258.1 | |
| AF532597.1 | 9 | 1137 | AAN33041.1 | |
| AK044188.1 | 9 | 1246 | BAC31809.1 | |
| AK089416.1 | 7 | 845 | BAC40873.1 | |
| AK154561.1 | 9 | 1233 | BAE32678.1 | |
| AK155821.1 | 9 | 1257 | ||
| AK155937.1 | 8 | 999 | BAE33511.1 | |
| AK157985.1 | 7 | 864 | ||
| BC050941.1 | 9 | 1146 | AAH50941.1 | |
| CA318154.1 | 3 | 280 |
More Information
Set Membership: |
|

Genomic Map