Assay Details
Target Gene Details
Entrez Gene ID: | 58200 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 1A |
Gene Aliases: |
0610038N18Rik, I-1 |
Location: |
Chr.15:103530279-103538005 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r1a | NM_021391.3 | 7 | 970 | NP_067366.1 |
| AK002813.1 | 3 | 604 | BAB22378.1 | |
| BC025123.1 | 7 | 939 | AAH25123.1 | |
| W75893.1 |
More Information
Set Membership: |
|

Genomic Map