Assay Details
Target Gene Details
Entrez Gene ID: | 26942 |
Gene Name: | sperm associated antigen 1 |
Gene Aliases: |
tpis |
Location: |
Chr.15:36175483-36235621 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spag1 | NM_012031.3 | 18 | 2566 | NP_036161.2 |
| AF181252.1 | 18 | 2558 | AAF06160.1 | |
| AF181253.1 | 8 | 1250 | AAF06161.1 | |
| AK032601.1 | 15 | 2361 | BAC27944.1 | |
| BC046313.1 | 18 | 2566 | AAH46313.1 |

Genomic Map