Assay Details
Target Gene Details
Entrez Gene ID: | 66497 |
Gene Name: | cms small ribosomal subunit 1 |
Gene Aliases: |
1110001A06Rik, 2610528E23Rik, 4930572F24Rik, AV099812, AV122629 |
Location: |
Chr.16:57302000-57606867 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cmss1 | NM_025599.3 | 10 | 930 | NP_079875.2 |
| AK012168.1 | 10 | 911 | BAB28074.1 | |
| AK019979.1 | 10 | 906 | BAB31948.1 | |
| BC038891.1 | 10 | 817 | AAH38891.1 |
More Information
Set Membership: |
|

Genomic Map