Assay Details
Target Gene Details
Entrez Gene ID: | 19065 |
Gene Name: | neuropeptide Y receptor Y4 |
Gene Aliases: |
NYYR-D, Ppyr1, Y4 |
Location: |
Chr.14:34145646-34152419 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npy4r | NM_008919.4 | 2 | 1610 | NP_032945.3 |
| BC132395.1 | 2 | 1610 | ||
| BC145981.1 | 2 | 1610 |
More Information
Set Membership: |
|

Genomic Map