Assay Details
Target Gene Details
Entrez Gene ID: | 223780 |
Gene Name: | adrenomedullin 2 |
Gene Aliases: |
Am2, IMD, Imdn |
Location: |
Chr.15:89322899-89324731 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adm2 | NM_182928.4 | 3 | 735 | NP_891558.1 |
| AB121035.1 | 3 | 659 | BAD07412.1 | |
| BB705319.1 | 1 | 114 | ||
| BC096428.1 | 3 | 662 | AAH96428.1 | |
| DV649647.1 | 3 | 611 |
More Information
Set Membership: |
|

Genomic Map