Assay Details
Target Gene Details
Entrez Gene ID: | 16539 |
Gene Name: | K+ voltage-gated channel, subfamily S, 2 |
Gene Aliases: |
E130006J24Rik, Kv9.2 |
Location: |
Chr.15:34837355-34843407 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcns2 | NM_001271704.1 | 2 | 772 | NP_001258633.1 |
| NM_181317.4 | 2 | 1140 | NP_851834.1 | |
| AK048819.1 | 2 | 1114 | BAC33468.1 | |
| AK087481.1 | 2 | 820 | BAC39892.1 | |
| AK173110.1 | 2 | 1140 | BAD32388.1 | |
| BC059833.1 | 2 | 772 | AAH59833.1 |
More Information
Set Membership: |
|

Genomic Map