Assay Details
Target Gene Details
Entrez Gene ID: | 76338 |
Gene Name: | RAB2B, member RAS oncogene family |
Gene Aliases: |
1500012D09Rik, 4930528G15Rik, A230002G14, D530043M21Rik |
Location: |
Chr.14:52261759-52279395 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rab2b | NM_172601.3 | 8 | 1379 | NP_766189.1 |
| AK005230.1 | BAB23894.1 | |||
| AK038393.1 | 8 | 1314 | BAC29983.1 | |
| AK169657.1 | 7 | 1254 | BAE41281.1 | |
| BC033312.1 | 8 | 1276 | AAH33312.1 | |
| BC046334.1 | 8 | 1321 | AAH46334.1 | |
| BC061513.1 | 3 | 2209 | AAH61513.1 |

Genomic Map