Assay Details
Target Gene Details
Entrez Gene ID: | 380959 |
Gene Name: | asparagine-linked glycosylation 10B (alpha-1,2-glucosyltransferase) |
Gene Aliases: |
AA469671, AU045404, AW123895, C85569, Deaf1, Gm917, nse5 |
Location: |
Chr.15:90224311-90230554 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alg10b | NM_001033441.3 | 1 | 239 | NP_001028613.1 |
| AK143038.1 | 1 | 245 | BAE25257.1 | |
| AK147821.1 | 1 | 240 | BAE28159.1 | |
| AK153580.1 | 1 | 219 | ||
| AK166344.1 | 1 | 245 | BAE38718.1 | |
| AK170450.1 | 1 | 240 | ||
| AK171179.1 | 1 | 201 |
More Information
Set Membership: |
|

Genomic Map