Assay Details
Target Gene Details
Entrez Gene ID: | 17988 |
Gene Name: | N-myc downstream regulated gene 1 |
Gene Aliases: |
CAP43, CMT4D, DRG1, HMSNL, NMSL, Ndr1, Ndrl, PROXY1, RTP, TDD5 |
Location: |
Chr.15:66929318-66969641 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndrg1 | NM_008681.2 | 16 | 1998 | NP_032707.2 |
| AK004548.1 | 16 | 1998 | BAB23362.1 | |
| AK098075.1 | 16 | 1997 | ||
| AK170439.1 | 16 | 1997 | BAE41797.1 | |
| AK170636.1 | 16 | 1997 | BAE41927.1 | |
| AK170809.1 | 16 | 1998 | BAE42042.1 | |
| BC015282.1 | 16 | 1953 | AAH15282.1 | |
| BC071235.1 | 16 | 2006 | AAH71235.1 | |
| U52073.1 | 17 | 1938 | AAB58249.1 | |
| U60593.1 | 16 | 1996 | AAB03484.1 |
More Information
Set Membership: |
|

Genomic Map