Assay Details
Target Gene Details
Entrez Gene ID: | 79456 |
Gene Name: | RecQ protein-like 4 |
Gene Aliases: |
RecQ4 |
Location: |
Chr.15:76703553-76710576 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Recql4 | NM_058214.3 | 15 | 2480 | NP_478121.2 |
| AB039882.1 | 15 | 2353 | BAD11131.1 | |
| AB175741.1 | 15 | 2397 | BAD14289.1 | |
| AK090260.1 | 16 | 2085 | ||
| BC131656.1 | 15 | 2393 |
More Information
Set Membership: |
|

Genomic Map