Assay Details
Target Gene Details
Entrez Gene ID: | 70646 |
Gene Name: | N(alpha)-acetyltransferase 30, NatC catalytic subunit |
Gene Aliases: |
4930487N19Rik, 5730533P17Rik, AI447922, AW322455, Nat12 |
Location: |
Chr.14:49172133-49191031 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Naa30 | NM_001081430.1 | 2 | 682 | NP_001074899.1 |
| AK015640.1 | 2 | 662 | BAC25468.1 | |
| AK044235.1 | 1 | 308 | ||
| BC157931.1 | 2 | 581 | ||
| BC158065.1 | 2 | 581 | ||
| BC172029.1 | 2 | 581 |
More Information
Set Membership: |
|

Genomic Map