Assay Details
Target Gene Details
Entrez Gene ID: | 68839 |
Gene Name: | ankyrin repeat domain 46 |
Gene Aliases: |
1110054N06Rik, AI314978, AI987733 |
Location: |
Chr.15:36477668-36503426 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd46 | NM_175134.4 | 3 | 417 | NP_780343.1 |
| AB076382.1 | 3 | 412 | BAD23946.1 | |
| AK079270.1 | BAC37594.1 | |||
| AK088304.1 | 3 | 378 | BAC40271.1 | |
| AK148049.1 | 3 | 378 | BAE28312.1 | |
| AK186948.1 | 1 | 176 | ||
| BC086659.1 | 3 | 384 | AAH86659.1 | |
| BC094595.1 | 3 | 380 | AAH94595.1 | |
| BQ715954.1 | 3 | 417 |

Genomic Map