Assay Details
Target Gene Details
Entrez Gene ID: | 223690 |
Gene Name: | ankyrin repeat domain 54 |
Gene Aliases: |
C730048E16Rik, EST1068184, EST475269, Liar |
Location: |
Chr.15:79053094-79062958 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd54 | NM_144849.1 | 8 | 1059 | NP_659098.1 |
| AK050440.1 | 7 | 1774 | ||
| AK133785.1 | 8 | 1083 | BAE21840.1 | |
| AK143625.1 | 8 | 1086 | BAE25471.1 | |
| BC014743.1 | 8 | 1059 | AAH14743.1 | |
| DQ884878.1 | 8 | 1087 |

Genomic Map