Assay Details
Target Gene Details
Entrez Gene ID: | 21847 |
Gene Name: | Kruppel-like factor 10 |
Gene Aliases: |
AI115143, EGR[a], Egral, Gdnfif, TIEG-1, Tieg, Tieg1, mGIF |
Location: |
Chr.15:38291464-38300711 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf10 | NM_001289471.1 | 3 | 995 | NP_001276400.1 |
| NM_013692.3 | 3 | 995 | NP_038720.1 | |
| AF049879.1 | 3 | 991 | AAC77796.1 | |
| AF064088.1 | 3 | 987 | AAC16734.1 | |
| AK004906.1 | 3 | 994 | ||
| AK043433.1 | 3 | 991 | BAC31547.1 | |
| AK079679.1 | 3 | 991 | ||
| AK145676.1 | 3 | 867 | ||
| AK154719.1 | 3 | 991 | BAE32785.1 | |
| BC003316.1 | 3 | 935 | AAH03316.1 | |
| Z36270.1 | 3 | 993 | CAA85283.1 |
More Information
Set Membership: |
|

Genomic Map