Assay Details
Target Gene Details
Entrez Gene ID: | 67747 |
Gene Name: | RIB43A domain with coiled-coils 2 |
Gene Aliases: |
4930579A10Rik, Trib |
Location: |
Chr.15:85132060-85144569 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ribc2 | NM_026357.2 | 7 | 1366 | NP_080633.2 |
| AK016311.1 | 7 | 1367 | BAB30190.2 | |
| AK017028.1 | 7 | 1385 | ||
| BC112373.1 | 7 | 1291 | AAI12374.1 |
More Information
Set Membership: |
|

Genomic Map