Assay Details
Target Gene Details
Entrez Gene ID: | 18383 |
Gene Name: | tumor necrosis factor receptor superfamily, member 11b (osteoprotegerin) |
Gene Aliases: |
OCIF, Opg, TR1 |
Location: |
Chr.15:54250619-54278484 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfrsf11b | NM_008764.3 | 2 | 333 | NP_032790.3 |
| AB013898.1 | 2 | 333 | BAA28269.1 | |
| AK140244.1 | 2 | 156 | BAE24295.1 | |
| AK146105.1 | 2 | 276 | BAE26905.1 | |
| BC049782.1 | 2 | 150 | AAH49782.1 | |
| U94331.1 | 2 | 186 | AAB53708.1 |
More Information
Set Membership: |
|

Genomic Map