Assay Details
Target Gene Details
Entrez Gene ID: | 224139 |
Gene Name: | golgi autoantigen, golgin subfamily b, macrogolgin 1 |
Gene Aliases: |
4930428L02Rik, 628101, 6330407A06Rik, AU042952, C130074L01Rik, F730017E11Rik, Gm6840, mKIAA4151 |
Location: |
Chr.16:36875173-36933085 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Golgb1 | NM_030035.1 | 13 | 6054 | NP_084311.1 |
| AK134596.1 | 1 | 42 | BAE22199.1 | |
| AK220188.1 | 1 | 1909 | BAD90373.1 | |
| BC145221.1 | ||||
| BC150731.1 | 13 | 6031 |

Genomic Map