Assay Details
Target Gene Details
Entrez Gene ID: | 75602 |
Gene Name: | RIKEN cDNA 1810062O18 gene |
Gene Aliases: |
- |
Location: |
Chr.14:20546293-20570680 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1810062O18Rik | NR_033571.1 | |||
| AK007938.1 | BAB25363.1 |
Target Gene Details
Entrez Gene ID: | 78787 |
Gene Name: | ubiquitin specific peptidase 54 |
Gene Aliases: |
4930429G18Rik, AI115571, C030002J06Rik |
Location: |
Chr.14:20548912-20643129 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp54 | NM_030180.2 | 18 | 4172 | NP_084456.2 |
| AK019588.1 | 1 | 359 | BAB31805.1 | |
| BC066166.1 | 1 | 683 | AAH66166.1 | |
| BC066175.1 | 1 | 683 | AAH66175.1 | |
| BC117844.1 | 1 | 347 | ||
| BC117845.1 | 1 | 347 |
More Information
Set Membership: |
|

Genomic Map