Assay Details
Target Gene Details
Entrez Gene ID: | 71939 |
Gene Name: | apolipoprotein L 6 |
Gene Aliases: |
2310076O14Rik |
Location: |
Chr.15:77044734-77057108 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apol6 | NM_001163621.1 | 4 | 578 | NP_001157093.1 |
| NM_028010.1 | 4 | 546 | NP_082286.1 | |
| AK010208.1 | 4 | 547 | BAB26769.1 | |
| AK144563.1 | 4 | 579 | BAE25940.1 |
More Information
Set Membership: |
|

Genomic Map