Assay Details
Target Gene Details
Entrez Gene ID: | 57259 |
Gene Name: | transducer of ERBB2, 2 |
Gene Aliases: |
2900090N22Rik, 4930545K18Rik, AV071822, mKIAA1663 |
Location: |
Chr.15:81848270-81858326 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tob2 | NM_020507.3 | 1 | 139 | NP_065253.1 |
| AK013833.1 | 2 | 348 | BAB29009.1 | |
| AK037631.1 | 1 | 136 | ||
| AK044623.1 | 1 | 137 | ||
| AK049766.1 | 1 | 86 | ||
| AK080817.1 | 1 | 87 | BAC38032.1 | |
| AK143542.1 | 1 | 116 | BAE25428.1 | |
| BC094031.1 | 1 | 70 | AAH94031.1 | |
| CK618657.1 | 1 | 251 |

Genomic Map