Assay Details
Target Gene Details
Entrez Gene ID: | 674895 |
Gene Name: | NIMA (never in mitosis gene a)- related kinase 10 |
Gene Aliases: |
Gm282 |
Location: |
Chr.14:14803580-15012059 on Build GRCm38 |
Assay Gene Location: | Within Intron 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nek10 | NM_001195229.1 | NP_001182158.1 | ||
| AK147862.1 | 11 | 2281 | BAE28185.1 |
More Information
Set Membership: |
|

Genomic Map