Assay Details
Target Gene Details
Entrez Gene ID: | 68705 |
Gene Name: | general transcription factor IIF, polypeptide 2 |
Gene Aliases: |
1110031C13Rik, AI326315 |
Location: |
Chr.14:75896937-76010865 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf2f2 | NM_026816.3 | NP_081092.1 | ||
| AK144858.1 | BAE26104.1 | |||
| BC027173.1 | AAH27173.1 |
Target Gene Details
Entrez Gene ID: | 67516 |
Gene Name: | potassium channel tetramerisation domain containing 4 |
Gene Aliases: |
2210017A09Rik, AU017169 |
Location: |
Chr.14:75955003-75965217 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kctd4 | NM_026214.3 | 2 | 1137 | NP_080490.3 |
| AK008741.1 | 2 | 1132 | BAB25870.1 | |
| AK017592.1 | 1 | 868 | BAB30826.1 | |
| AK032314.1 | 1 | 844 | BAC27807.1 | |
| AK032624.1 | 1 | 875 | BAC27956.1 | |
| BC028748.1 | 1 | 852 | AAH28748.1 |
More Information
Set Membership: |
|

Genomic Map